Assay Details
Target Gene Details
Entrez Gene ID: | 653602 |
Gene Name: | uncharacterized LOC653602 |
Gene Aliases: |
- |
Location: |
Chr.2:14635091-14650819 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC653602 | NM_001291410.1 | NP_001278339.1 | ||
| AK092711.1 | BAC03955.1 |

Genomic Map