Assay Details
Target Gene Details
Entrez Gene ID: | 51011 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 2A |
Gene Aliases: |
CGI-105 |
Location: |
Chr.2:95402691-95416702 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| esv2759076 | Chr.2:95304390 - 95486811 on Build GRCh38 | Loss |
|
| nsv998014 | Chr.2:95247530 - 95630882 on Build GRCh38 | Gain |
|
| nsv961449 | Chr.2:95403429 - 95415505 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map