Assay Details
Target Gene Details
Entrez Gene ID: | 64225 |
Gene Name: | atlastin GTPase 2 |
Gene Aliases: |
ARL3IP2, ARL6IP2, atlastin2 |
Location: |
Chr.2:38293957-38378579 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATL2 | NM_001135673.2 | NP_001129145.1 | ||
| NM_001308076.1 | NP_001295005.1 | |||
| NM_022374.3 | NP_071769.2 | |||
| NR_024191.1 | ||||
| XM_011533017.1 | XP_011531319.1 | |||
| XM_011533018.2 | XP_011531320.1 | |||
| XM_011533019.2 | XP_011531321.1 | |||
| XM_011533020.1 | XP_011531322.1 | |||
| XM_011533022.2 | XP_011531324.1 | |||
| XM_011533023.2 | XP_011531325.1 | |||
| XM_017004686.1 | XP_016860175.1 | |||
| XM_017004687.1 | XP_016860176.1 | |||
| XM_017004688.1 | XP_016860177.1 | |||
| AK026946.1 | BAB15598.1 | |||
| AK225190.1 | ||||
| AK294049.1 | ||||
| AK294711.1 | ||||
| AK302621.1 | ||||
| AK303985.1 | ||||
| AK308894.1 | ||||
| BC053508.1 | AAH53508.1 | |||
| BX507348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map