Assay Details
Target Gene Details
Entrez Gene ID: | 9168 |
Gene Name: | thymosin beta 10 |
Gene Aliases: |
MIG12, TB10 |
Location: |
Chr.2:84905639-84906675 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMSB10 | NM_021103.3 | NP_066926.1 | ||
| AK130949.1 | ||||
| AK311952.1 | ||||
| AY453400.1 | AAS47517.1 | |||
| BC016025.1 | AAH16025.1 | |||
| BC016731.1 | AAH16731.1 | |||
| M20259.1 | AAA36744.1 | |||
| M92381.1 | AAC41691.1 | |||
| S54005.1 | AAB25225.1 |
More Information
Set Membership: |
|

Genomic Map