Assay Details
Target Gene Details
Entrez Gene ID: | 389072 |
Gene Name: | pleckstrin homology domain containing M3 |
Gene Aliases: |
DAPR, PLEKHM1L |
Location: |
Chr.2:207821288-208025675 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHM3 | NM_001080475.2 | 8 | 8545 | NP_001073944.1 |
| XM_005246545.3 | 7 | 8387 | XP_005246602.1 | |
| XM_011511160.2 | 9 | 8880 | XP_011509462.1 | |
| XM_017004072.1 | 7 | 8730 | XP_016859561.1 | |
| XM_017004073.1 | 7 | 7609 | XP_016859562.1 | |
| AF462444.1 | 1 | 225 | AAP97712.1 | |
| BX538248.1 | 1 | 581 |
More Information
Set Membership: |
|

Genomic Map