Assay Details
Target Gene Details
Entrez Gene ID: | 93349 |
Gene Name: | SP140 nuclear body protein like |
Gene Aliases: |
- |
Location: |
Chr.2:230327152-230407506 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP140L | NM_001308162.1 | NP_001295091.1 | ||
| NM_001308163.1 | NP_001295092.1 | |||
| NM_138402.4 | NP_612411.4 | |||
| XM_006712855.2 | XP_006712918.1 | |||
| XM_006712856.2 | XP_006712919.1 | |||
| XM_006712858.3 | XP_006712921.1 | |||
| XM_011512190.2 | XP_011510492.1 | |||
| XM_011512191.1 | XP_011510493.1 | |||
| XM_017005294.1 | XP_016860783.1 | |||
| XM_017005295.1 | XP_016860784.1 | |||
| XM_017005296.1 | XP_016860785.1 | |||
| XM_017005297.1 | XP_016860786.1 | |||
| XM_017005298.1 | XP_016860787.1 | |||
| XM_017005299.1 | XP_016860788.1 | |||
| XM_017005300.1 | XP_016860789.1 | |||
| XM_017005301.1 | XP_016860790.1 | |||
| BC004921.2 | AAH04921.1 | |||
| BX281497.1 | ||||
| DB240458.1 | ||||
| KF419365.1 | ||||
| KF419366.1 | ||||
| KF419367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29172 | Chr.2:230208263 - 230603132 on Build GRCh38 | Loss |
|
| nsv584667 | Chr.2:230313107 - 230393735 on Build GRCh38 | Loss |
|
| dgv4180n100 | Chr.2:230309705 - 230393888 on Build GRCh38 | Loss |
|
| esv2721594 | Chr.2:230295549 - 230405436 on Build GRCh38 | Deletion |
|
| nsv1001929 | Chr.2:230315169 - 230393888 on Build GRCh38 | Gain+Loss |
|
| esv3594601 | Chr.2:230326507 - 230364470 on Build GRCh38 | Loss |
|
| esv3594600 | Chr.2:230286077 - 230385943 on Build GRCh38 | Loss |
|
| dgv7334n54 | Chr.2:230314161 - 230393735 on Build GRCh38 | Gain |
|
| esv3893565 | Chr.2:230318606 - 230393735 on Build GRCh38 | Gain+Loss |
|
| esv2751844 | Chr.2:230314780 - 230415780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map