Assay Details
Target Gene Details
Entrez Gene ID: | 79823 |
Gene Name: | calmodulin-lysine N-methyltransferase |
Gene Aliases: |
C2orf34, CLNMT, CaM KMT, Cam, KMT |
Location: |
Chr.2:44361714-44774388 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMKMT | NM_024766.4 | NP_079042.1 | ||
| XM_011533111.2 | XP_011531413.1 | |||
| XM_017004970.1 | XP_016860459.1 | |||
| XM_017004971.1 | XP_016860460.1 | |||
| XM_017004972.1 | XP_016860461.1 | |||
| XM_017004973.1 | XP_016860462.1 | |||
| XM_017004974.1 | XP_016860463.1 | |||
| XM_017004975.1 | XP_016860464.1 | |||
| XM_017004976.1 | XP_016860465.1 | |||
| XM_017004977.1 | XP_016860466.1 | |||
| XM_017004978.1 | XP_016860467.1 | |||
| XM_017004979.1 | XP_016860468.1 | |||
| XM_017004980.1 | XP_016860469.1 | |||
| XM_017004981.1 | XP_016860470.1 | |||
| AK027104.1 | BAB15658.1 | |||
| BC029359.1 | AAH29359.1 | |||
| BC053733.1 | AAH53733.1 | |||
| DA760277.1 | ||||
| DB461740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3798n100 | Chr.2:44279994 - 44404126 on Build GRCh38 | Loss |
|
| nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
|
| esv3583770 | Chr.2:44369957 - 44383099 on Build GRCh38 | Loss |
|
| esv2762539 | Chr.2:44357175 - 44447311 on Build GRCh38 | Loss |
|
| esv3892026 | Chr.2:44318437 - 44418772 on Build GRCh38 | Loss |
|
| nsv833947 | Chr.2:44341139 - 44515256 on Build GRCh38 | Gain |
|
| nsv581698 | Chr.2:44349410 - 44407077 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map