Assay Details
Target Gene Details
Entrez Gene ID: | 102724058 |
Gene Name: | uncharacterized LOC102724058 |
Gene Aliases: |
- |
Location: |
Chr.2:165957418-166036400 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724058 | NR_110598.1 | |||
| HY008183.1 | ||||
| HY011842.1 | ||||
| HY029566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834445 | Chr.2:165800387 - 165995152 on Build GRCh38 | Gain+Loss |
|

Genomic Map