Assay Details
Target Gene Details
Entrez Gene ID: | 731220 |
Gene Name: | RFX family member 8, lacking RFX DNA binding domain |
Gene Aliases: |
- |
Location: |
Chr.2:101397344-101475179 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX8 | NM_001145664.1 | NP_001139136.1 | ||
| XM_011511771.2 | XP_011510073.1 | |||
| XM_011511772.1 | XP_011510074.1 | |||
| XM_011511773.1 | XP_011510075.1 | |||
| XM_011511776.1 | XP_011510078.1 | |||
| XM_011511777.1 | XP_011510079.1 | |||
| XM_011511778.1 | XP_011510080.1 | |||
| XM_017004851.1 | XP_016860340.1 | |||
| XM_017004852.1 | XP_016860341.1 | |||
| XM_017004853.1 | XP_016860342.1 | |||
| AK124976.1 | BAC86013.1 | |||
| AK131289.1 | BAD18461.1 | |||
| AK298610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892993 | Chr.2:101281409 - 101650795 on Build GRCh38 | Gain |
|
| esv3591767 | Chr.2:101402309 - 101533995 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map