Assay Details
Target Gene Details
Entrez Gene ID: | 5899 |
Gene Name: | RALB Ras like proto-oncogene B |
Gene Aliases: |
- |
Location: |
Chr.2:120240070-120294711 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALB | NM_002881.2 | NP_002872.1 | ||
| XM_005263724.1 | XP_005263781.1 | |||
| XM_005263727.1 | XP_005263784.1 | |||
| XM_005263728.1 | XP_005263785.1 | |||
| XM_005263729.2 | XP_005263786.1 | |||
| XM_011511573.1 | XP_011509875.1 | |||
| XM_011511574.1 | XP_011509876.1 | |||
| XM_017004622.1 | XP_016860111.1 | |||
| AK127675.1 | BAC87080.1 | |||
| AK303214.1 | ||||
| AK304588.1 | ||||
| BC018163.1 | AAH18163.1 | |||
| M35416.1 | AAA60250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000477 | Chr.2:120103257 - 120287829 on Build GRCh38 | Gain |
|
| nsv508164 | Chr.2:120196044 - 120296224 on Build GRCh38 | Deletion |
|
| nsv834346 | Chr.2:120132786 - 120304522 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map