Assay Details
Target Gene Details
Entrez Gene ID: | 51281 |
Gene Name: | ankyrin repeat and MYND domain containing 1 |
Gene Aliases: |
ZMYND13 |
Location: |
Chr.2:240468632-240561110 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKMY1 | XM_017004271.1 | XP_016859760.1 | ||
| XM_017004272.1 | XP_016859761.1 | |||
| XM_017004273.1 | XP_016859762.1 | |||
| XM_017004274.1 | XP_016859763.1 | |||
| XM_017004275.1 | XP_016859764.1 | |||
| XM_017004276.1 | XP_016859765.1 | |||
| XM_017004277.1 | XP_016859766.1 | |||
| XM_017004278.1 | XP_016859767.1 | |||
| XM_017004279.1 | XP_016859768.1 | |||
| XM_017004281.1 | XP_016859770.1 | |||
| XM_017004282.1 | XP_016859771.1 | |||
| XM_017004284.1 | XP_016859773.1 | |||
| XM_017004285.1 | XP_016859774.1 | |||
| XM_017004288.1 | XP_016859777.1 | |||
| XM_017004290.1 | XP_016859779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759119 | Chr.2:240425704 - 240649454 on Build GRCh38 | Gain |
|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
| nsv508899 | Chr.2:240333579 - 240490606 on Build GRCh38 | Insertion |
|
| esv3594799 | Chr.2:240471618 - 240478176 on Build GRCh38 | Gain |
|
| nsv834586 | Chr.2:240475181 - 240622625 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map