Assay Details
Target Gene Details
Entrez Gene ID: | 84141 |
Gene Name: | eva-1 homolog A, regulator of programmed cell death |
Gene Aliases: |
FAM176A, TMEM166 |
Location: |
Chr.2:75492318-75569722 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVA1A | NM_001135032.1 | NP_001128504.1 | ||
| NM_032181.2 | NP_115557.1 | |||
| AK023453.1 | BAB14580.1 | |||
| BC016157.1 | AAH16157.1 | |||
| BC063016.1 | AAH63016.1 | |||
| BE892427.1 | ||||
| BP276065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759063 | Chr.2:75462279 - 75785697 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map