Assay Details
Target Gene Details
Entrez Gene ID: | 151507 |
Gene Name: | male-specific lethal 3 homolog (Drosophila) pseudogene 1 |
Gene Aliases: |
MSL3L2 |
Location: |
Chr.2:233865437-233868409 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSL3P1 | NR_024322.1 | |||
| AK301707.1 | ||||
| AK316372.1 | ||||
| BI831020.1 | ||||
| DB457661.1 | ||||
| DC394932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013712 | Chr.2:233649811 - 234587181 on Build GRCh38 | Loss |
|
| nsv3216 | Chr.2:233864792 - 233896078 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map