Assay Details
Target Gene Details
Entrez Gene ID: | 189 |
Gene Name: | alanine-glyoxylate aminotransferase |
Gene Aliases: |
AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6 |
Location: |
Chr.2:240868745-240879119 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGXT | NM_000030.2 | NP_000021.1 | ||
| AB292648.1 | ||||
| AF348451.1 | AAK30157.1 | |||
| AK292754.1 | ||||
| AK298400.1 | ||||
| AK298420.1 | ||||
| AK315331.1 | ||||
| AK316495.1 | ||||
| BC132819.1 | ||||
| D13368.1 | BAA02632.1 | |||
| X53414.1 | CAA37493.1 | |||
| X56092.1 | CAA39572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584861 | Chr.2:240810727 - 241018260 on Build GRCh38 | Gain |
|
| nsv955294 | Chr.2:240710484 - 240925283 on Build GRCh38 | Deletion |
|
| nsv470531 | Chr.2:240684476 - 240928657 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map