Assay Details
Target Gene Details
Entrez Gene ID: | 440900 |
Gene Name: | long intergenic non-protein coding RNA 1191 |
Gene Aliases: |
VIN, lnc-ACTR3 |
Location: |
Chr.2:113979569-114007310 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01191 | NR_034128.1 |

Genomic Map