Assay Details
Target Gene Details
Entrez Gene ID: | 729723 |
Gene Name: | DNAJC27 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:24972112-25039694 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC27-AS1 | NR_034113.1 | |||
| CR933670.1 | ||||
| DA314471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004529 | Chr.2:24363242 - 25005468 on Build GRCh38 | Loss |
|
| nsv1008732 | Chr.2:24907673 - 25349661 on Build GRCh38 | Loss |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|

Genomic Map