Assay Details
Target Gene Details
Entrez Gene ID: | 10890 |
Gene Name: | RAB10, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.2:26033860-26137454 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB10 | NM_016131.4 | NP_057215.3 | ||
| AF086917.1 | AAP97147.1 | |||
| AF091032.1 | AAF00045.1 | |||
| AF106681.1 | AAD43034.1 | |||
| AF297660.1 | AAG13413.1 | |||
| AF498945.1 | AAM21093.1 | |||
| AK023223.1 | BAB14474.1 | |||
| AL136650.1 | CAB66585.1 | |||
| AL517754.3 | ||||
| BC000896.1 | AAH00896.1 | |||
| CR457303.1 | CAG33584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508717 | Chr.2:26043125 - 26131976 on Build GRCh38 | Deletion |
|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map