Assay Details
Target Gene Details
Entrez Gene ID: | 402117 |
Gene Name: | von Willebrand factor C domain containing protein 2-like |
Gene Aliases: |
- |
Location: |
Chr.2:214411737-214575929 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWC2L | NM_001080500.2 | 2 | 410 | NP_001073969.1 |
| AB374231.1 | 1 | 280 | ||
| BC146903.1 | 2 | 410 | ||
| BC146911.1 | 2 | 410 | ||
| BC146931.1 | 2 | 410 | ||
| BC146935.1 | 2 | 410 | ||
| BC171841.1 | 2 | 410 | ||
| EF552207.1 | 1 | 283 | ||
| EU541473.1 | 1 | 283 |
More Information
Set Membership: |
|

Genomic Map