Assay Details
Target Gene Details
Entrez Gene ID: | 52 |
Gene Name: | acid phosphatase 1, soluble |
Gene Aliases: |
HAAP, LMW-PTP |
Location: |
Chr.2:264869-278283 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP1 | NM_001040649.2 | NP_001035739.1 | ||
| NM_004300.3 | NP_004291.1 | |||
| NM_007099.3 | NP_009030.1 | |||
| NR_024080.1 | ||||
| AB209838.1 | BAD93075.1 | |||
| AK289934.1 | ||||
| AK291861.1 | ||||
| BC007422.2 | AAH07422.1 | |||
| BC106011.1 | AAI06012.1 | |||
| BI460080.1 | ||||
| BJ992018.1 | ||||
| BT007136.1 | AAP35800.1 | |||
| DA378283.1 | ||||
| DB443811.1 | ||||
| M83653.1 | AAB59354.1 | |||
| M83654.1 | AAB59355.1 | |||
| Y16846.1 | CAA76416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589599 | Chr.2:251988 - 283726 on Build GRCh38 | Gain |
|
| esv3589598 | Chr.2:250064 - 298533 on Build GRCh38 | Gain |
|
| nsv833159 | Chr.2:158491 - 327294 on Build GRCh38 | Loss |
|
| esv2759016 | Chr.2:167949 - 929902 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map