Assay Details
Target Gene Details
Entrez Gene ID: | 55437 |
Gene Name: | STE20-related kinase adaptor beta |
Gene Aliases: |
ALS2CR2, CALS-21, ILPIP, ILPIPA, PAPK, PRO1038 |
Location: |
Chr.2:201451669-201480851 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRADB | NM_001206864.1 | NP_001193793.1 | ||
| NM_018571.5 | NP_061041.2 | |||
| XM_005246669.1 | XP_005246726.1 | |||
| XM_017004438.1 | XP_016859927.1 | |||
| AB038950.1 | BAB32500.1 | |||
| AK027637.1 | BAB55254.1 | |||
| AK290457.1 | ||||
| AY093697.1 | AAM19143.1 | |||
| BC008302.1 | AAH08302.1 | |||
| BC090871.1 | AAH90871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575325 | Chr.2:201441625 - 201498896 on Build GRCh38 | Gain |
|
| nsv3111 | Chr.2:201458871 - 201493181 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map