Assay Details
Target Gene Details
Entrez Gene ID: | 307 |
Gene Name: | annexin A4 |
Gene Aliases: |
ANX4, HEL-S-274, P32.5, PAP-II, PIG28, PP4-X, ZAP36 |
Location: |
Chr.2:69643805-69826477 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANXA4 | NM_001320698.1 | NP_001307627.1 | ||
| XM_017003943.1 | XP_016859432.1 | |||
| XM_017003944.1 | XP_016859433.1 | |||
| DN994496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33940 | Chr.2:69115288 - 69789041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map