Assay Details
Target Gene Details
Entrez Gene ID: | 400960 |
Gene Name: | PCBP1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:69962263-70087015 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP1-AS1 | NR_033872.1 | 11 | 1315 | |
| AK308634.1 | 3 | 410 |

Genomic Map