Assay Details
Target Gene Details
Entrez Gene ID: | 84141 |
Gene Name: | eva-1 homolog A, regulator of programmed cell death |
Gene Aliases: |
FAM176A, TMEM166 |
Location: |
Chr.2:75492318-75569722 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVA1A | NM_001135032.1 | NP_001128504.1 | ||
| NM_032181.2 | NP_115557.1 | |||
| AK023453.1 | BAB14580.1 | |||
| BC016157.1 | AAH16157.1 | |||
| BC063016.1 | AAH63016.1 | |||
| BE892427.1 | ||||
| CR457314.1 | CAG33595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759063 | Chr.2:75462279 - 75785697 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116403185] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map