Assay Details
Target Gene Details
Entrez Gene ID: | 284958 |
Gene Name: | 5'-nucleotidase domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.2:112717245-112742879 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5DC4 | XM_017005477.1 | XP_016860966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954792 | Chr.2:112709124 - 112728823 on Build GRCh38 | Deletion |
|
| nsv526301 | Chr.2:112693921 - 112738208 on Build GRCh38 | Loss |
|
| nsv441775 | Chr.2:112704822 - 112721310 on Build GRCh38 | Loss |
|
| esv2665843 | Chr.2:112702882 - 112727074 on Build GRCh38 | Deletion |
|
| esv2759084 | Chr.2:112711658 - 112721312 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map