Assay Details
Target Gene Details
Entrez Gene ID: | 150681 |
Gene Name: | olfactory receptor family 6 subfamily B member 3 |
Gene Aliases: |
OR6B3P, OR6B3Q |
Location: |
Chr.2:240044611-240053722 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6B3 | XM_011510671.2 | XP_011508973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
| esv3594791 | Chr.2:240042983 - 240049904 on Build GRCh38 | Loss |
|
| nsv3236 | Chr.2:240037431 - 240067330 on Build GRCh38 | Insertion |
|
| nsv525278 | Chr.2:240001219 - 240122324 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78672179] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map