Assay Details
Target Gene Details
Entrez Gene ID: | 729475 |
Gene Name: | RAD51 associated protein 2 |
Gene Aliases: |
- |
Location: |
Chr.2:17510584-17530414 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51AP2 | NM_001099218.2 | 3 | 3409 | NP_001092688.1 |
| NM_001321233.1 | 7 | 3767 | NP_001308162.1 | |
| XM_011533084.1 | 2 | 3328 | XP_011531386.1 | |
| AK310498.1 | 8 | 4014 | ||
| BG216055.1 | 2 | 144 | ||
| DB514225.1 | 2 | 114 | ||
| DQ860102.1 | 3 | 3409 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997502 | Chr.2:17160707 - 18147073 on Build GRCh38 | Gain |
|

Genomic Map