Assay Details
Target Gene Details
Entrez Gene ID: | 8708 |
Gene Name: | beta-1,3-galactosyltransferase 1 |
Gene Aliases: |
beta3Gal-T1 |
Location: |
Chr.2:167293060-167870856 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALT1 | XM_005246931.3 | XP_005246988.1 | ||
| XM_011512085.2 | XP_011510387.1 | |||
| E07739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008704 | Chr.2:167297015 - 167339861 on Build GRCh38 | Gain |
|
| esv3584156 | Chr.2:167306613 - 167332085 on Build GRCh38 | Loss |
|
| esv2763488 | Chr.2:167306626 - 167324930 on Build GRCh38 | Loss |
|
| nsv3020 | Chr.2:167287266 - 167320708 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map