Assay Details
Target Gene Details
Entrez Gene ID: | 7881 |
Gene Name: | potassium voltage-gated channel subfamily A member regulatory beta subunit 1 |
Gene Aliases: |
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3 |
Location: |
Chr.3:156118211-156540603 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNAB1 | XM_011513115.2 | 14 | 4112 | XP_011511417.1 |
| XM_011513117.2 | 12 | 4002 | XP_011511419.1 | |
| XM_017007171.1 | 14 | 4189 | XP_016862660.1 | |
| XM_017007172.1 | 14 | 5405 | XP_016862661.1 | |
| XM_017007173.1 | 12 | 4106 | XP_016862662.1 |
Target Gene Details
Entrez Gene ID: | 6747 |
Gene Name: | signal sequence receptor subunit 3 |
Gene Aliases: |
TRAPG |
Location: |
Chr.3:156539553-156555200 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSR3 | NM_001308197.1 | 5 | 4089 | NP_001295126.1 |
| NM_001308204.1 | 5 | 3915 | NP_001295133.1 | |
| NM_001308205.1 | 5 | 3939 | NP_001295134.1 | |
| NM_007107.4 | 5 | 4050 | NP_009038.1 | |
| AI225105.1 | 1 | 282 | ||
| BJ997360.1 | 1 | 375 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829764 | Chr.3:156457035 - 156629985 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map