Assay Details
Target Gene Details
Entrez Gene ID: | 200844 |
Gene Name: | chromosome 3 open reading frame 67 |
Gene Aliases: |
- |
Location: |
Chr.3:58716415-59050078 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf67 | NM_198463.2 | NP_940865.1 | ||
| XM_005264929.2 | XP_005264986.1 | |||
| XM_006713013.3 | XP_006713076.1 | |||
| XM_011533451.2 | XP_011531753.1 | |||
| XM_011533453.2 | XP_011531755.1 | |||
| XM_011533455.2 | XP_011531757.1 | |||
| XM_011533456.1 | XP_011531758.1 | |||
| XM_011533457.1 | XP_011531759.1 | |||
| XM_011533458.2 | XP_011531760.1 | |||
| XM_011533459.2 | XP_011531761.1 | |||
| XM_011533460.2 | XP_011531762.1 | |||
| XM_011533461.1 | XP_011531763.1 | |||
| XM_011533463.2 | XP_011531765.1 | |||
| XM_017005848.1 | XP_016861337.1 | |||
| XM_017005849.1 | XP_016861338.1 | |||
| XM_017005850.1 | XP_016861339.1 | |||
| AK124111.1 | BAC85775.1 | |||
| AK124920.1 | BAC85994.1 | |||
| BC050317.1 | AAH50317.1 | |||
| BC132815.1 | ||||
| BC151142.1 |
Target Gene Details
Entrez Gene ID: | 101929238 |
Gene Name: | C3orf67 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:58824471-59019093 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf67-AS1 | NR_110820.1 | |||
| AK090895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain |
|
| nsv997365 | Chr.3:58848323 - 58930257 on Build GRCh38 | Loss |
|

Genomic Map