Assay Details
Target Gene Details
Entrez Gene ID: | 253461 |
Gene Name: | zinc finger and BTB domain containing 38 |
Gene Aliases: |
CIBZ, PPP1R171, ZNF921 |
Location: |
Chr.3:141324213-141449792 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB38 | NM_001080412.2 | NP_001073881.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2678380 | Chr.3:141351255 - 141356724 on Build GRCh38 | Deletion |
|
| esv3562635 | Chr.3:141351159 - 141356743 on Build GRCh38 | Deletion |
|
| nsv829744 | Chr.3:141297907 - 141470326 on Build GRCh38 | Loss |
|
| nsv460871 | Chr.3:141308772 - 141356120 on Build GRCh38 | Loss |
|
| esv3597976 | Chr.3:141351206 - 141356749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map