Assay Details
Target Gene Details
Entrez Gene ID: | 23218 |
Gene Name: | neurobeachin like 2 |
Gene Aliases: |
BDPLT4, GPS |
Location: |
Chr.3:46979683-47009704 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBEAL2 | NM_015175.2 | NP_055990.1 | ||
| XM_006713072.3 | XP_006713135.1 | |||
| XM_011533533.2 | XP_011531835.1 | |||
| XM_017006010.1 | XP_016861499.1 | |||
| XM_017006011.1 | XP_016861500.1 | |||
| XM_017006012.1 | XP_016861501.1 | |||
| XM_017006013.1 | XP_016861502.1 | |||
| XM_017006014.1 | XP_016861503.1 | |||
| XM_017006015.1 | XP_016861504.1 | |||
| XM_017006016.1 | 1 | 242 | XP_016861505.1 | |
| AK092871.1 | BAC03994.1 | |||
| AK131104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954479 | Chr.3:46975211 - 47013210 on Build GRCh38 | Deletion |
|
| nsv590209 | Chr.3:46976724 - 47009294 on Build GRCh38 | Loss |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
| esv3596073 | Chr.3:46936296 - 47131646 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map