Assay Details
Target Gene Details
Entrez Gene ID: | 57633 |
Gene Name: | leucine rich repeat neuronal 1 |
Gene Aliases: |
FIGLER3, NLRR-1 |
Location: |
Chr.3:3799431-3849834 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRN1 | NM_001324188.1 | NP_001311117.1 | ||
| NM_001324189.1 | NP_001311118.1 | |||
| NM_020873.6 | NP_065924.3 | |||
| AK291717.1 | ||||
| BC034947.2 | AAH34947.1 | |||
| DA736067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000460 | Chr.3:3399757 - 3993339 on Build GRCh38 | Gain |
|
| nsv1011650 | Chr.3:3797741 - 3934721 on Build GRCh38 | Loss |
|
| esv3893637 | Chr.3:3512223 - 3972206 on Build GRCh38 | Loss |
|
| nsv236590 | Chr.3:3800684 - 3800759 on Build GRCh38 | Deletion |
|
| esv2724816 | Chr.3:3800019 - 3801007 on Build GRCh38 | Deletion |
|
| esv2752001 | Chr.3:3539056 - 3858107 on Build GRCh38 | Gain |
|
| nsv589396 | Chr.3:3203825 - 3821595 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map