Assay Details
Target Gene Details
Entrez Gene ID: | 10777 |
Gene Name: | cAMP regulated phosphoprotein 21 |
Gene Aliases: |
ARPP-21, R3HDM3, RCS, TARPP |
Location: |
Chr.3:35638981-35798933 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPP21 | NM_001267616.1 | 1 | 551 | NP_001254545.1 |
| XM_017005578.1 | XP_016861067.1 | |||
| BC041385.1 | 1 | 139 | AAH41385.1 |
More Information
Set Membership: |
|

Genomic Map