Assay Details
Target Gene Details
Entrez Gene ID: | 1370 |
Gene Name: | carboxypeptidase N subunit 2 |
Gene Aliases: |
ACBP |
Location: |
Chr.3:194339765-194351387 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPN2 | NM_001080513.3 | NP_001073982.3 | ||
| NM_001291988.1 | NP_001278917.1 | |||
| XM_005269280.4 | XP_005269337.1 | |||
| BC031569.1 | ||||
| BC042334.1 | ||||
| BC137398.1 | ||||
| BC137403.1 | ||||
| BX951477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2563733 | Chr.3:194347576 - 194348690 on Build GRCh38 | Insertion |
|
| dgv2671n106 | Chr.3:194347571 - 194348271 on Build GRCh38 | Deletion |
|
| nsv1073274 | Chr.3:194347570 - 194348471 on Build GRCh38 | Deletion |
|
| esv24336 | Chr.3:194347350 - 194348384 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111683075] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map