Assay Details
Target Gene Details
Entrez Gene ID: | 389158 |
Gene Name: | phospholipid scramblase family member 5 |
Gene Aliases: |
- |
Location: |
Chr.3:146576555-146606216 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLSCR5 | NM_001085420.1 | NP_001078889.1 | ||
| NM_001321245.1 | NP_001308174.1 | |||
| XM_017006373.1 | XP_016861862.1 | |||
| AY436642.1 | ||||
| BC157886.1 | ||||
| BC171917.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72988660] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map