Assay Details
Target Gene Details
Entrez Gene ID: | 6747 |
Gene Name: | signal sequence receptor subunit 3 |
Gene Aliases: |
TRAPG |
Location: |
Chr.3:156539553-156555200 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSR3 | NM_001308197.1 | NP_001295126.1 | ||
| NM_001308204.1 | NP_001295133.1 | |||
| NM_001308205.1 | NP_001295134.1 | |||
| NM_007107.4 | NP_009038.1 | |||
| AF087907.1 | AAP97205.1 | |||
| AF110647.1 | AAD48587.1 | |||
| AK304364.1 | ||||
| AK312935.1 | ||||
| BC017203.1 | AAH17203.1 | |||
| BT006669.1 | AAP35315.1 | |||
| DA135338.1 | ||||
| DA283706.1 | ||||
| DA725890.1 | ||||
| DA750309.1 | ||||
| HY035536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829764 | Chr.3:156457035 - 156629985 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map