Assay Details
Target Gene Details
Entrez Gene ID: | 152217 |
Gene Name: | NCBP2 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:196942623-196944013 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCBP2-AS2 | NR_024388.1 | 1 | 722 | |
| AK123020.1 | 1 | 671 | ||
| AL832915.1 | 1 | 658 | CAH10618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| nsv593086 | Chr.3:196897131 - 196958250 on Build GRCh38 | Loss |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|

Genomic Map