Assay Details
Target Gene Details
Entrez Gene ID: | 344807 |
Gene Name: | CD200 receptor 1 like |
Gene Aliases: |
CD200R2, CD200RLa |
Location: |
Chr.3:112814925-112847004 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD200R1L | XM_011512763.2 | 8 | 2177 | XP_011511065.1 |
| XM_017006300.1 | 7 | 1554 | XP_016861789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597421 | Chr.3:112807512 - 112904954 on Build GRCh38 | Gain |
|
| nsv591293 | Chr.3:112266260 - 113293950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map