Assay Details
Target Gene Details
Entrez Gene ID: | 846 |
Gene Name: | calcium sensing receptor |
Gene Aliases: |
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1 |
Location: |
Chr.3:122183683-122286503 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASR | NM_000388.3 | NP_000379.2 | ||
| NM_001178065.1 | NP_001171536.1 | |||
| XM_006713789.3 | XP_006713852.1 | |||
| U20759.1 | AAA86503.1 | |||
| U20760.1 | AAA86504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591402 | Chr.3:122170241 - 122215314 on Build GRCh38 | Loss |
|
| esv33139 | Chr.3:121781553 - 122372984 on Build GRCh38 | Loss |
|
| dgv8526n54 | Chr.3:122181001 - 122216999 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139879325] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map