Assay Details
Target Gene Details
Entrez Gene ID: | 7123 |
Gene Name: | C-type lectin domain family 3 member B |
Gene Aliases: |
TN, TNA |
Location: |
Chr.3:45026207-45036073 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC3B | NM_001308394.1 | NP_001295323.1 | ||
| NM_003278.2 | NP_003269.2 | |||
| XM_017007116.1 | XP_016862605.1 | |||
| AK312093.1 | ||||
| BC011024.1 | AAH11024.1 | |||
| BI826174.1 | ||||
| BI826652.1 | ||||
| BI837909.1 | ||||
| CD616729.1 | ||||
| CR541981.1 | CAG46778.1 | |||
| CR542009.1 | CAG46806.1 | |||
| CR749292.1 | 3 | 769 | CAH18147.1 | |
| X64559.1 | CAA45860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508914 | Chr.3:45004646 - 45047422 on Build GRCh38 | Insertion |
|
| esv2343097 | Chr.3:45031660 - 45032079 on Build GRCh38 | Deletion |
|
| nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
|
| esv2725216 | Chr.3:45031342 - 45032234 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map