Assay Details
Target Gene Details
Entrez Gene ID: | 2317 |
Gene Name: | filamin B |
Gene Aliases: |
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP |
Location: |
Chr.3:58008400-58172255 on Build GRCh38 |
Assay Gene Location: | Within Intron 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLNB | NM_001164317.1 | NP_001157789.1 | ||
| NM_001164318.1 | NP_001157790.1 | |||
| NM_001164319.1 | NP_001157791.1 | |||
| NM_001457.3 | NP_001448.2 | |||
| XM_005264978.1 | XP_005265035.1 | |||
| AB191258.1 | BAD52434.1 | |||
| AB209889.1 | BAD93126.1 | |||
| AB371580.1 | ||||
| AB371581.1 | ||||
| AB371582.1 | ||||
| AF042166.1 | AAC39842.1 | |||
| AF043045.1 | AAC33845.1 | |||
| AF238609.1 | AAF97046.1 | |||
| AL137574.1 | CAB70818.1 | |||
| AL833551.1 | ||||
| BX641085.1 | CAE46040.1 | |||
| CR749793.1 | CAH18654.1 | |||
| M62994.1 | AAA35505.1 |
Target Gene Details
Entrez Gene ID: | 105377105 |
Gene Name: | uncharacterized LOC105377105 |
Gene Aliases: |
- |
Location: |
Chr.3:58162547-58170635 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377105 | NR_135534.1 | |||
| DB066566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834703 | Chr.3:58072791 - 58298770 on Build GRCh38 | Gain |
|
| nsv528147 | Chr.3:57929747 - 58384450 on Build GRCh38 | Gain |
|
| nsv1001814 | Chr.3:58158881 - 58354197 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75163724] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map