Assay Details
Target Gene Details
Entrez Gene ID: | 1370 |
Gene Name: | carboxypeptidase N subunit 2 |
Gene Aliases: |
ACBP |
Location: |
Chr.3:194339765-194351387 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPN2 | NM_001080513.3 | NP_001073982.3 | ||
| NM_001291988.1 | NP_001278917.1 | |||
| XM_005269280.4 | XP_005269337.1 | |||
| BC031569.1 | ||||
| BC042334.1 | ||||
| BC137398.1 | ||||
| BC137403.1 | ||||
| BX951477.1 |
More Information
Set Membership: |
|

Genomic Map