Assay Details
Target Gene Details
Entrez Gene ID: | 55831 |
Gene Name: | ER membrane protein complex subunit 3 |
Gene Aliases: |
POB, TMEM111 |
Location: |
Chr.3:9962680-10011248 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC3 | NM_018447.3 | NP_060917.1 | ||
| XM_005265321.3 | XP_005265378.1 | |||
| DA359966.1 |
Target Gene Details
Entrez Gene ID: | 442075 |
Gene Name: | EMC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:9986893-10007004 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC3-AS1 | NR_103821.1 | |||
| AK092352.1 | BAC03867.1 | |||
| AK125558.1 | ||||
| DR004716.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013567 | Chr.3:9990575 - 10084686 on Build GRCh38 | Loss |
|
| nsv966988 | Chr.3:9993448 - 9996549 on Build GRCh38 | Duplication |
|
| esv2724891 | Chr.3:9988492 - 10047899 on Build GRCh38 | Deletion |
|
| nsv589648 | Chr.3:9892921 - 10042251 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map