Assay Details
Target Gene Details
Entrez Gene ID: | 1370 |
Gene Name: | carboxypeptidase N subunit 2 |
Gene Aliases: |
ACBP |
Location: |
Chr.3:194339765-194351387 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPN2 | NM_001080513.3 | 2 | 860 | NP_001073982.3 |
| NM_001291988.1 | 2 | 857 | NP_001278917.1 | |
| XM_005269280.4 | 3 | 989 | XP_005269337.1 | |
| BC031569.1 | 2 | 808 | ||
| BC042334.1 | 2 | 816 | ||
| BC137398.1 | 2 | 850 | ||
| BC137403.1 | 2 | 850 | ||
| J05158.2 | 1 | 774 | AAA51921.2 |
More Information
Set Membership: |
|

Genomic Map