Assay Details
Target Gene Details
Entrez Gene ID: | 55840 |
Gene Name: | ELL associated factor 2 |
Gene Aliases: |
BM040, TRAITS, U19 |
Location: |
Chr.3:121835180-121886526 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EAF2 | NM_001320041.1 | NP_001306970.1 | ||
| NM_018456.5 | NP_060926.2 | |||
| XM_005247618.3 | 2 | 266 | XP_005247675.1 | |
| XM_017006861.1 | XP_016862350.1 | |||
| XM_017006862.1 | XP_016862351.1 | |||
| XM_017006863.1 | XP_016862352.1 | |||
| AF217516.1 | AAF67627.1 | |||
| AF517829.1 | AAO63811.1 | |||
| AK301560.1 | ||||
| AY049020.1 | AAL12223.1 | |||
| BC014209.2 | AAH14209.1 | |||
| HY005912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain |
|
| nsv516233 | Chr.3:121700454 - 121951558 on Build GRCh38 | Gain+Loss |
|
| esv33139 | Chr.3:121781553 - 122372984 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map