Assay Details
Target Gene Details
Entrez Gene ID: | 100128640 |
Gene Name: | ACVR2B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:38451027-38454820 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR2B-AS1 | NR_028389.1 | |||
| AK025619.1 |

Genomic Map