Assay Details
Target Gene Details
Entrez Gene ID: | 7104 |
Gene Name: | transmembrane 4 L six family member 4 |
Gene Aliases: |
ILTMP, il-TMP |
Location: |
Chr.3:149474581-149503394 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF4 | NM_004617.3 | NP_004608.1 | ||
| AK055577.1 | ||||
| AK315464.1 | ||||
| BC001386.2 | AAH01386.1 | |||
| BT007430.1 | AAP36098.1 | |||
| U31449.1 | AAA74745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29983 | Chr.3:149433515 - 149484515 on Build GRCh38 | Gain |
|
| nsv4056 | Chr.3:149441396 - 149485702 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77207040] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map