Assay Details
Target Gene Details
Entrez Gene ID: | 103344926 |
Gene Name: | uncharacterized LincR-0002 |
Gene Aliases: |
- |
Location: |
Chr.3:191425524-191590356 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINCR-0002 | NR_120606.1 | |||
| BU620307.1 |
Target Gene Details
Entrez Gene ID: | 152138 |
Gene Name: | pyrin domain containing 2 |
Gene Aliases: |
POP2, cPOP2 |
Location: |
Chr.3:191461163-191461456 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYDC2 | NM_001083308.1 | 1 | 178 | NP_001076777.1 |
| AY858112.1 | 1 | 178 | AAX54598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428428 | Chr.3:191219045 - 191497807 on Build GRCh38 | Gain |
|
| esv3599095 | Chr.3:191251629 - 191553437 on Build GRCh38 | Loss |
|
| esv2759209 | Chr.3:191149222 - 191561318 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map