Assay Details
Target Gene Details
Entrez Gene ID: | 10777 |
Gene Name: | cAMP regulated phosphoprotein 21 |
Gene Aliases: |
ARPP-21, R3HDM3, RCS, TARPP |
Location: |
Chr.3:35638981-35798933 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
More Information
Set Membership: |
|

Genomic Map