Assay Details
Target Gene Details
Entrez Gene ID: | 1173 |
Gene Name: | adaptor related protein complex 2 mu 1 subunit |
Gene Aliases: |
AP50, CLAPM1, mu2 |
Location: |
Chr.3:184174846-184184091 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP2M1 | NM_001025205.1 | NP_001020376.1 | ||
| NM_001311198.1 | NP_001298127.1 | |||
| NM_004068.3 | NP_004059.2 | |||
| AK057883.1 | ||||
| AK294159.1 | ||||
| AK295999.1 | ||||
| AK297578.1 | ||||
| AK297849.1 | ||||
| AK304510.1 | ||||
| BC004996.1 | AAH04996.1 | |||
| BC013796.1 | AAH13796.1 | |||
| BC014030.1 | AAH14030.1 | |||
| BI461415.1 | ||||
| D63475.1 | ||||
| DB261680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map